Assay Details
| Gene Symbol: Arid4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194262.2 | NP_919238.1 | 15 - 16 | 1703 | 159 | |
| NM_198122.2 | NP_937755.1 | 15 - 16 | 1703 | 159 | ||
| GenBank mRNA | AK020165.1 | - | 1 - 2 | 101 | 159 | |
| AK031062.1 | - | 15 - 16 | 1686 | 159 | ||
| AK080355.1 | - | 5 - 6 | 563 | 159 | ||
| AK136105.1 | - | 15 - 16 | 1587 | 159 | ||
| BC137783.1 | - | 14 - 15 | 1465 | 8860802 | 159 | |
| CB522383.1 | - | 1 - 2 | 118 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arid4b
Genomic Map