Assay Details
| Gene Symbol: Stat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019963.1 | NP_064347.1 | 11 - 12 | 1248 | 60 | |
| GenBank mRNA | AF187231.1 | - | 11 - 12 | 1242 | 60 | |
| AF206162.1 | - | 11 - 12 | 1248 | 60 | ||
| AK030903.1 | - | 4 - 5 | 340 | 60 | ||
| AK149924.1 | - | 11 - 12 | 1261 | 60 | ||
| AK150070.1 | - | 11 - 12 | 1260 | 60 | ||
| AK153405.1 | - | 9 - 10 | 886 | 60 | ||
More Information
Set Membership: |
|

Stat2
Genomic Map