Assay Details
| Gene Symbol: Tcaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203396.1 | NP_981933.1 | 3 - 4 | 1779 | 78 | |
| GenBank mRNA | AY528666.1 | - | 3 - 4 | 1779 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcaf3
Genomic Map