Assay Details
| Gene Symbol: Tmevpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104123.1 | 1 - 2 | 70 | 83 | ||
| GenBank mRNA | BX519358.1 | - | 1 - 2 | 70 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interferon gamma | Ifng |
IFN-g, Ifg |
-79.2kb | Anti-sense | Chr. 10 - 118441047 - 118445892 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 33337 |
Gm33337 | 158.0kb | Sense | Chr. 10 - 118714522 - 118751241 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmevpg1
Genomic Map