Assay Details
| Gene Symbol: Mylk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166030.1 | NP_001159502.1 | 5 - 6 | 616 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mylk4| Gene Symbol: Mylk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166030.1 | NP_001159502.1 | 5 - 6 | 616 | 93 | |
Set Membership: |
|