Assay Details
| Gene Symbol: Wrnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030215.3 | NP_084491.3 | 2 - 3 | 1209 | 126 | |
| GenBank mRNA | AB056151.1 | - | 2 - 3 | 1058 | 126 | |
| AF208046.1 | - | 2 - 3 | 454 | 126 | ||
| AK041886.1 | - | 2 - 3 | 1165 | 126 | ||
| AK050368.1 | - | 2 - 3 | 1211 | 126 | ||
| AK082078.1 | - | 2 - 3 | 1124 | 126 | ||
| AK167570.1 | - | 2 - 3 | 1210 | 126 | ||
| AK170542.1 | - | 2 - 3 | 1202 | 126 | ||
| AK170846.1 | - | 2 - 3 | 1190 | 126 | ||
| BC010482.1 | - | 2 - 3 | 1201 | 3492125 | 126 | |
| BC058744.1 | - | 2 - 3 | 599 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wrnip1
Genomic Map