Assay Details
| Gene Symbol: Wrnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030215.3 | NP_084491.3 | 5 - 6 | 1831 | 64 | |
| GenBank mRNA | AB056151.1 | - | 5 - 6 | 1680 | 64 | |
| AF208046.1 | - | 5 - 6 | 1076 | 64 | ||
| AK041886.1 | - | 5 - 6 | 1787 | 64 | ||
| AK050368.1 | - | 5 - 6 | 1833 | 64 | ||
| AK167570.1 | - | 5 - 6 | 1832 | 64 | ||
| AK170542.1 | - | 5 - 6 | 1824 | 64 | ||
| AK170846.1 | - | 5 - 6 | 1812 | 64 | ||
| AK205588.1 | - | 1 - 2 | 69 | 64 | ||
| AK209893.1 | - | 2 - 3 | 180 | 64 | ||
| BC058744.1 | - | 5 - 6 | 1221 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wrnip1
Genomic Map