Assay Details
| Gene Symbol: Galnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144908.3 | NP_659157.1 | 6 - 7 | 1142 | 149 | |
| GenBank mRNA | AF495528.1 | - | 5 - 6 | 1014 | 149 | |
| AK088014.1 | - | 6 - 7 | 1131 | 149 | ||
| AK161524.1 | - | 6 - 7 | 1052 | 149 | ||
| BC011428.1 | - | 6 - 7 | 1118 | 2631696 | 149 | |
| BC021504.1 | - | 6 - 7 | 1143 | 5346678 | 149 | |
| BC031150.1 | - | 5 - 6 | 1040 | 149 | ||
| BC034184.1 | - | 5 - 6 | 1042 | 4974216 | 149 | |
| BC034185.1 | - | 5 - 6 | 1094 | 4974695 | 149 | |
| BC036143.1 | - | 5 - 6 | 1043 | 4974645 | 149 | |
| BC036145.1 | - | 5 - 6 | 1036 | 4975474 | 149 | |
| Y12435.1 | - | 5 - 6 | 965 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Galnt11
Genomic Map