Assay Details
| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294162.1 | NP_001281091.1 | 1 - 2 | 430 | 75 | |
| GenBank mRNA | AF012869.1 | - | 1 - 2 | 378 | 75 | |
| AF034762.1 | - | 1 - 2 | 422 | 75 | ||
| AK145127.1 | - | 431 | 75 | |||
More Information
Set Membership: |
|

Kcnh2
Genomic Map