Assay Details
| Gene Symbol: Smim10l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271583.1 | NP_001258512.1 | 1 - 2 | 369 | 65 | |
| GenBank mRNA | BY708088.1 | - | 1 - 2 | 353 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smim10l1
Genomic Map