Assay Details
| Gene Symbol: Smim10l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073368.1 | 2 - 3 | 526 | 89 | ||
| GenBank mRNA | AA048533.1 | - | 2 - 3 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim10l1| Gene Symbol: Smim10l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073368.1 | 2 - 3 | 526 | 89 | ||
| GenBank mRNA | AA048533.1 | - | 2 - 3 | 89 | ||
Set Membership: |
|