Assay Details
| Gene Symbol: Prl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163530.1 | NP_001157002.1 | 1 - 2 | 90 | 85 | |
| GenBank mRNA | AK017520.1 | - | 1 - 2 | 86 | 85 | |
| AK017584.1 | - | 1 - 2 | 84 | 85 | ||
| AK133645.1 | - | 1 - 2 | 84 | 85 | ||
| AK161553.1 | - | 1 - 2 | 83 | 85 | ||
| AK161594.1 | - | 1 - 2 | 84 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017520.1, AK017584.1, AK133645.1, AK161553.1, AK161594.1, NM_001163530.1 transcripts.
Set Membership: |
|

Prl
Genomic Map