Assay Details
| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023729.3 | NP_076218.3 | 6 - 7 | 711 | 102 | |
| GenBank mRNA | AF459789.1 | - | 6 - 7 | 705 | 102 | |
| AK016595.1 | - | 6 - 7 | 706 | 102 | ||
More Information
Set Membership: |
|

Asz1
Genomic Map