Assay Details
| Gene Symbol: Foxp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286607.1 | NP_001273536.1 | 2 - 3 | 490 | 77 | |
| NM_053242.4 | NP_444472.2 | 4 - 5 | 827 | 77 | ||
| NM_212435.1 | NP_997600.1 | 2 - 3 | 490 | 77 | ||
| GenBank mRNA | AF339106.1 | - | 1 - 2 | 168 | 77 | |
| AK051668.1 | - | 4 - 5 | 789 | 77 | ||
| AK082361.1 | - | 4 - 5 | 499 | 77 | ||
| AY079003.1 | - | 4 - 5 | 456 | 77 | ||
| BC058960.1 | - | 2 - 3 | 487 | 6811877 | 77 | |
| BC062926.1 | - | 2 - 3 | 490 | 6851153 | 77 | |
More Information
Set Membership: |
|

Foxp2
Genomic Map