Assay Details
Gene Symbol: Tsga13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054073.2 | NP_473414.1 | 2 - 3 | 248 | 61 | |
GenBank mRNA | AF294348.1 | - | 2 - 3 | 216 | 61 | |
AK006265.1 | - | 2 - 3 | 249 | 61 | ||
BC125597.1 | - | 1 - 2 | 166 | 40130112 | 61 | |
BC125599.1 | - | 1 - 2 | 166 | 40130114 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF294348.1, AK006265.1, BC125597.1, BC125599.1, NM_054073.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
