Assay Details
| Gene Symbol: Tac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC144737.1 | - | 4 - 5 | 324 | 9053285 | 71 |
| M68909.1 | - | 3 - 4 | 145 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Tac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC144737.1 | - | 4 - 5 | 324 | 9053285 | 71 |
| M68909.1 | - | 3 - 4 | 145 | 71 | ||
Set Membership: |
|