Assay Details
| Gene Symbol: Mest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252292.1 | NP_001239221.1 | 2 - 3 | 383 | 60 | |
| GenBank mRNA | AK146457.1 | - | 2 - 3 | 383 | 60 | |
More Information
Set Membership: |
|

Mest
Genomic Map