Assay Details
| Gene Symbol: Zfp184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183014.1 | NP_898835.1 | 4 - 5 | 548 | 126 | |
| GenBank mRNA | BC053084.1 | - | 4 - 5 | 548 | 5708591 | 126 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp184
Genomic Map