| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000022888.mm.1 |
NM_001163024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022888.mm.1 |
NM_001163024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022888.mm.1 |
NM_001163025.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022888.mm.1 |
NM_001163025.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022888.mm.1 |
NM_153395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022888.mm.1 |
NM_153395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022888.mm.1 |
TC1000003066.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000022889.mm.1 |
NM_001163024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022889.mm.1 |
NM_001163024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022889.mm.1 |
NM_001163025.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022889.mm.1 |
NM_001163025.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022889.mm.1 |
NM_153395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022889.mm.1 |
NM_153395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022889.mm.1 |
TC1000003066.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000022887.mm.1 |
NM_001163024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022887.mm.1 |
NM_001163024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022887.mm.1 |
NM_001163025.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022887.mm.1 |
NM_001163025.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022887.mm.1 |
NM_153395.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022887.mm.1 |
NM_153395.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000022887.mm.1 |
TC1000003066.mm.1 |
MTA TC |