Assay Details
| Gene Symbol: 1700025F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040578.1 | 1 - 2 | 172 | 119 | ||
| GenBank mRNA | AK015773.1 | - | 1 - 2 | 151 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

1700025F24Rik
Genomic Map