Assay Details
| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172280.2 | NP_758484.2 | 10 - 11 | 1536 | 85 | |
| GenBank mRNA | BC039956.1 | - | 10 - 11 | 1527 | 85 | |
| BC066050.1 | - | 8 - 9 | 1396 | 85 | ||
| BC089304.1 | - | 10 - 11 | 1536 | 6854147 | 85 | |
More Information
Set Membership: |
|

2210018M11Rik
Genomic Map