Assay Details
| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172280.2 | NP_758484.2 | 13 - 14 | 2018 | 91 | |
| GenBank mRNA | AK032985.1 | - | 12 - 13 | 2003 | 91 | |
| BC039956.1 | - | 13 - 14 | 2008 | 91 | ||
| BC066050.1 | - | 11 - 12 | 1878 | 91 | ||
| BC089304.1 | - | 13 - 14 | 2018 | 6854147 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

2210018M11Rik
Genomic Map