Assay Details
| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172280.2 | NP_758484.2 | 2 - 3 | 203 | 69 | |
| GenBank mRNA | AK032985.1 | - | 2 - 3 | 230 | 69 | |
| AK137691.1 | - | 2 - 3 | 419 | 69 | ||
| BC039956.1 | - | 2 - 3 | 194 | 69 | ||
| BC066050.1 | - | 2 - 3 | 180 | 69 | ||
| BC089304.1 | - | 2 - 3 | 203 | 6854147 | 69 | |
More Information
Set Membership: |
|

2210018M11Rik
Genomic Map