Assay Details
| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172280.2 | NP_758484.2 | 9 - 10 | 1385 | 75 | |
| GenBank mRNA | AK032985.1 | - | 8 - 9 | 1370 | 75 | |
| BC039956.1 | - | 9 - 10 | 1376 | 75 | ||
| BC066050.1 | - | 7 - 8 | 1245 | 75 | ||
| BC089304.1 | - | 9 - 10 | 1385 | 6854147 | 75 | |
More Information
Set Membership: |
|

2210018M11Rik
Genomic Map