Assay Details
| Gene Symbol: Pak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011035.2 | NP_035165.2 | 1 - 2 | 97 | ||
| GenBank mRNA | AK077497.1 | - | 1 - 2 | 97 | ||
| BC055740.1 | - | 1 - 2 | 97 | |||
| BC060157.1 | - | 1 - 2 | 97 | |||
| BC079838.1 | - | 1 - 2 | 97 | |||
| BC145698.1 | - | 1 - 3 | 127 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_011035.2 transcripts.
Set Membership: |
|

Pak1
Genomic Map