Assay Details
| Gene Symbol: Cdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007664.4 | NP_031690.3 | 11 - 12 | 2394 | 96 | |
| GenBank mRNA | AB008811.1 | - | 11 - 12 | 2071 | 96 | |
| AK032826.1 | - | 11 - 12 | 1834 | 96 | ||
| AK146981.1 | - | 11 - 12 | 2591 | 96 | ||
| BC022107.1 | - | 11 - 12 | 1825 | 5100313 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh2
Genomic Map