Assay Details
| Gene Symbol: Cdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007664.4 | NP_031690.3 | 7 - 8 | 1672 | 125 | |
| GenBank mRNA | AB008811.1 | - | 7 - 8 | 1349 | 125 | |
| AK032826.1 | - | 7 - 8 | 1112 | 125 | ||
| AK146981.1 | - | 7 - 8 | 1869 | 125 | ||
| BC022107.1 | - | 7 - 8 | 1103 | 5100313 | 125 | |
| M31131.1 | - | 7 - 8 | 1349 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh2
Genomic Map