Assay Details
Gene Symbol: Tcaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020704.1 | - | 2 - 3 | 222 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK020704.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
