Assay Details
| Gene Symbol: Msl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010832.5 | NP_034962.2 | 2 - 3 | 267 | 72 | |
| GenBank mRNA | AK089368.1 | - | 2 - 3 | 350 | 72 | |
| AK146208.1 | - | 2 - 3 | 204 | 72 | ||
| AK148898.1 | - | 2 - 3 | 246 | 72 | ||
| BC010226.1 | - | 2 - 3 | 228 | 3501126 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Msl3
Genomic Map