Assay Details
| Gene Symbol: Smo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176996.4 | NP_795970.3 | 9 - 10 | 2178 | 69 | |
| GenBank mRNA | AK081050.1 | - | 3 - 4 | 366 | 69 | |
| BC030377.1 | - | 2 - 3 | 215 | 69 | ||
| BC096028.1 | - | 9 - 10 | 1697 | 6850065 | 69 | |
| BQ947565.1 | - | 4 - 5 | 440 | 69 | ||
| FJ883552.1 | - | 9 - 10 | 1687 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smo
Genomic Map