Assay Details
| Gene Symbol: Zfp800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081678.1 | NP_001075147.1 | 1 - 2 | 123 | 99 | |
| GenBank mRNA | BC119236.1 | - | 1 - 2 | 123 | 8733985 | 99 |
| BC119262.1 | - | 1 - 2 | 123 | 8734011 | 99 | |
| BC144761.1 | - | 1 - 2 | 123 | 9053309 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC119236.1, BC119262.1, BC144761.1, NM_001081678.1 transcripts.
Set Membership: |
|

Zfp800
Genomic Map