Assay Details
| Gene Symbol: Svopl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177200.4 | NP_796174.2 | 10 - 11 | 971 | 69 | |
| GenBank mRNA | AK034997.1 | - | 10 - 11 | 972 | 69 | |
| AK137043.1 | - | 10 - 11 | 972 | 69 | ||
| BC058741.1 | - | 10 - 11 | 954 | 6309979 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Svopl
Genomic Map