Assay Details
| Gene Symbol: Svopl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177200.4 | NP_796174.2 | 11 - 12 | 1184 | 80 | |
| GenBank mRNA | AK078436.1 | - | 1 - 2 | 154 | 80 | |
| BC058741.1 | - | 11 - 12 | 1167 | 6309979 | 80 | |
More Information
Set Membership: |
|

Svopl
Genomic Map