Assay Details
| Gene Symbol: Svopl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177200.4 | NP_796174.2 | 1 - 2 | 76 | 72 | |
| GenBank mRNA | AK034997.1 | - | 1 - 2 | 77 | 72 | |
| AK137043.1 | - | 1 - 2 | 77 | 72 | ||
| BB627230.1 | - | 1 - 2 | 77 | 72 | ||
| BC058741.1 | - | 1 - 2 | 59 | 6309979 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034997.1, AK137043.1, BC058741.1, NM_177200.4 transcripts.
Set Membership: |
|

Svopl
Genomic Map