Assay Details
| Gene Symbol: Prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014651.1 | - | 1 - 2 | 290 | 106 | |
| AK029349.1 | - | 1 - 2 | 290 | 106 | ||
| AK165551.1 | - | 1 - 2 | 358 | 106 | ||
More Information
Set Membership: |
|

Prdx4
Genomic Map