Assay Details
| Gene Symbol: Galnt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173739.3 | NP_776100.2 | 2 - 3 | 864 | 76 | |
| GenBank mRNA | AB175685.1 | - | 2 - 3 | 434 | 76 | |
| BC024988.1 | - | 2 - 3 | 864 | 5044626 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt18
Genomic Map