Assay Details
| Gene Symbol: Galnt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173739.3 | NP_776100.2 | 7 - 8 | 1754 | 62 | |
| GenBank mRNA | AB175685.1 | - | 7 - 8 | 1279 | 62 | |
| AK207434.1 | - | 1 - 2 | 52 | 62 | ||
| BC024988.1 | - | 7 - 8 | 1754 | 5044626 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt18
Genomic Map