Assay Details
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027629.3 | NP_081905.1 | 5 - 6 | 872 | 70 | |
GenBank mRNA | AK016437.1 | - | 3 - 4 | 231 | 70 | |
AK039189.1 | - | 5 - 6 | 856 | 70 | ||
AK040513.1 | - | 5 - 6 | 792 | 70 | ||
BC020351.1 | - | 5 - 6 | 1085 | 70 | ||
BC032150.1 | - | 1 - 2 | 78 | 5004308 | 70 | |
BC076571.1 | - | 5 - 6 | 769 | 30356163 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
