| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600016950.mm.1 |
TC0600002147.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016950.mm.1 |
NM_028550.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016950.mm.1 |
NM_028550.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016949.mm.1 |
TC0600002147.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016949.mm.1 |
NM_028550.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016949.mm.1 |
NM_028550.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016952.mm.1 |
TC0600002147.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016952.mm.1 |
NM_028550.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016952.mm.1 |
NM_028550.3 |
Refseq |