Assay Details
| Gene Symbol: Abca16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278943.1 | NP_001265872.1 | 10 - 11 | 1555 | 92 | |
| NM_001278944.1 | NP_001265873.1 | 13 - 14 | 2024 | 92 | ||
| GenBank mRNA | AK144723.1 | - | 10 - 11 | 1556 | 92 | |
| AK161253.1 | - | 2 - 3 | 307 | 92 | ||
| AY243472.1 | - | 13 - 14 | 2023 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abca16
Genomic Map