Assay Details
| Gene Symbol: Abca16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278943.1 | NP_001265872.1 | 23 - 24 | 4022 | 64 | |
| NM_001278944.1 | NP_001265873.1 | 26 - 27 | 4491 | 64 | ||
| GenBank mRNA | AY243472.1 | - | 26 - 27 | 4490 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abca16
Genomic Map