Assay Details
| Gene Symbol: Abca16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278944.1 | NP_001265873.1 | 7 - 8 | 1110 | 139 | |
| GenBank mRNA | AY243472.1 | - | 7 - 8 | 1110 | 139 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278944.1 transcripts.
Set Membership: |
|

Abca16
Genomic Map