Assay Details
| Gene Symbol: Abca16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278943.1 | NP_001265872.1 | 5 - 6 | 812 | 82 | |
| GenBank mRNA | AK144723.1 | - | 5 - 6 | 813 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abca16
Genomic Map