Assay Details
| Gene Symbol: Abca15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177213.3 | NP_796187.2 | 13 - 14 | 2031 | 95 | |
| GenBank mRNA | AK029712.1 | - | 14 - 15 | 2032 | 95 | |
| AY243471.1 | - | 13 - 14 | 2031 | 95 | ||
| BC141350.1 | - | 13 - 14 | 1996 | 9055962 | 95 | |
| BC141351.1 | - | 13 - 14 | 1996 | 9055963 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abca15
Genomic Map