Assay Details
| Gene Symbol: Abca15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177213.3 | NP_796187.2 | 25 - 26 | 4242 | 89 | |
| GenBank mRNA | AY243471.1 | - | 25 - 26 | 4242 | 89 | |
| BC141350.1 | - | 25 - 26 | 4207 | 9055962 | 89 | |
| BC141351.1 | - | 25 - 26 | 4207 | 9055963 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abca15
Genomic Map