Assay Details
| Gene Symbol: Abca15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177213.3 | NP_796187.2 | 27 - 28 | 4627 | 64 | |
| GenBank mRNA | AY243471.1 | - | 27 - 28 | 4627 | 64 | |
| BC141350.1 | - | 27 - 28 | 4592 | 9055962 | 64 | |
| BC141351.1 | - | 27 - 28 | 4592 | 9055963 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abca15
Genomic Map