Assay Details
Gene Symbol: 4933427G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028955.1 | NP_083231.1 | 5 - 6 | 1405 | 103 | |
GenBank mRNA | AK016951.1 | - | 5 - 6 | 1405 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258848 | AK016951.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258848 | AK016951.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102341 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160229 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258848 | AK016951.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258848 | AK016951.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160229 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06346014 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102341 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06346044 | - | - |
Whole Mouse Genome G4122A | A_52_P324716 | NM_028955.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P324716 | AK016951.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258848 | AK016951.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258848 | AK016951.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114385 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102341 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471614 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06336425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471527 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06202123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06313677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06313678 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471608 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06637575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25471566 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06313680 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1431905_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431904_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431905_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1431904_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431905_s_at | - | - |
Mouse Genome 430 2.0 Array | 1431904_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74466_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74466_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700014797.mm.1 | TC0700001738.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700014797.mm.1 | NM_028955.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700014797.mm.1 | NM_028955.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700014796.mm.1 | TC0700001738.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700014796.mm.1 | NM_028955.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700014796.mm.1 | NM_028955.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700014798.mm.1 | TC0700001738.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700014798.mm.1 | NM_028955.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700014798.mm.1 | NM_028955.1 | Refseq |
Mouse Expression Set 430 | 1431904_PM_at | - | - |
Mouse Expression Set 430 | 1431905_s_at | - | - |
Mouse Expression Set 430 | 1431904_at | - | - |
Mouse Expression Set 430 | 1431905_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431904_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431904_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431905_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431905_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 783109 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2260487 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2260487 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002260487 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213416 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1213416 | - | - |
mouse-6 v1.1 (Illumina) | scl32112.9.4_161-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2260487 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32112.9.4_161-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260487 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775861 | - | - |
