Assay Details
| Gene Symbol: Ctbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170744.1 | NP_001164215.1 | 1 - 2 | 1726 | 125 | |
| GenBank mRNA | BC017520.1 | - | 2 - 3 | 1571 | 4511826 | 125 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC017520.1 transcripts.
Set Membership: |
|

Ctbp2
Genomic Map