Assay Details
| Gene Symbol: Adam8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089086.1 | - | 15 - 16 | 1643 | 66 | |
| AK134118.1 | - | 5 - 6 | 1197 | 66 | ||
| AK150411.1 | - | 15 - 16 | 1645 | 66 | ||
| AK151535.1 | - | 7 - 8 | 874 | 66 | ||
| AK152675.1 | - | 15 - 16 | 1662 | 66 | ||
| AK152769.1 | - | 15 - 16 | 1644 | 66 | ||
| AK153065.1 | - | 15 - 16 | 1644 | 66 | ||
| AK155772.1 | - | 15 - 16 | 1643 | 66 | ||
| AK155912.1 | - | 15 - 16 | 1663 | 66 | ||
| AK160041.1 | - | 15 - 16 | 1646 | 66 | ||
| AK170019.1 | - | 15 - 16 | 1668 | 66 | ||
| AK171060.1 | - | 15 - 16 | 1643 | 66 | ||
| BC025584.1 | - | 15 - 16 | 1697 | 5324943 | 66 | |
| X13335.1 | - | 15 - 16 | 1642 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam8
Genomic Map