Assay Details
| Gene Symbol: Tmem80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141950.1 | NP_001135422.1 | 2 - 3 | 282 | 71 | |
| GenBank mRNA | AK134821.1 | - | 2 - 3 | 283 | 71 | |
| AK169083.1 | - | 2 - 3 | 241 | 71 | ||
| BC022756.1 | - | 2 - 3 | 226 | 4922223 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK134821.1, AK169083.1, BC022756.1, NM_001141950.1 transcripts.
Set Membership: |
|

Tmem80
Genomic Map